This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03383313_u1
See other FCGRT GE Assays ›
Gene Symbol:
FCGRT
Gene Name:
Fc fragment of IgG receptor and transporter
Gene Aliases:
FcRn
Chromosome Location:
Chr.6: 54581103 - 54599193 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03383313_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FCGRT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214197.2 NP_999362.2 5 947 74
XM_013998635.2 XP_013854089.1 1093 74
XM_013998636.2 XP_013854090.1 1150 74
XM_013998637.2 XP_013854091.1 1108 74
XM_013998639.2 XP_013854093.1 1077 74
XM_021093552.1 XP_020949211.1 1189 74
XM_021093554.1 XP_020949213.1 1060 74
GenBank mRNA AK232709.1 - 5 1094 74
AK235834.1 - 6 1029 74
AK236469.1 - 5 1094 74
AK236505.1 - 6 1013 74
AK239628.1 - 6 1010 74
AK394503.1 - 6 989 74
AK394940.1 - 6 1066 74
AK395598.1 - 5 1140 74
AY244705.1 - 5 920 74
AY740682.1 - 5 947 74
EF208068.1 - 4 671 74
EU852582.1 - 3 584 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-q595s:80/100.66.78.76:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0