This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03383662_u1
See other GHR GE Assays ›
Gene Symbol:
GHR
Gene Name:
growth hormone receptor
Gene Aliases:
-
Chromosome Location:
Chr.16: 27126300 - 27422618 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03383662_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GHR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214254.2 NP_999419.2 9 1854 71
XM_013990636.2 XP_013846090.2 2438 71
XM_013990637.2 XP_013846091.2 2305 71
XM_021076567.1 XP_020932226.1 19479 71
XM_021076568.1 XP_020932227.1 1998 71
XM_021076569.1 XP_020932228.1 2444 71
XM_021076570.1 XP_020932229.1 1883 71
XM_021076571.1 XP_020932230.1 2371 71
XM_021076572.1 XP_020932231.1 2367 71
XM_021076573.1 XP_020932232.1 1884 71
XM_021076574.1 XP_020932233.1 1883 71
XM_021076575.1 XP_020932234.1 1866 71
GenBank mRNA AK233142.1 - 11 2077 71
DQ106869.1 - 9 1854 71
DQ422962.1 - 9 1854 71
EF041823.1 - 9 1854 71
HM013749.1 - 9 1865 71
JF276446.1 - 9 1854 71
KC999114.1 - 9 1854 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
cytokine-mediated signaling pathway
positive regulation of peptidyl-tyrosine phosphorylation
growth hormone receptor signaling pathway
cytokine receptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0