This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03383715_u1
See other IL1RN GE Assays ›
Gene Symbol:
IL1RN
Gene Name:
interleukin 1 receptor antagonist
Gene Aliases:
IRAP1
Chromosome Location:
-
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03383715_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL1RN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214262.1 NP_999427.1 454 70
GenBank mRNA AY577820.1 - 206 70
L38849.1 - 454 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Inventoried

Panther Classification:

Molecular Function -

interleukin superfamily

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cytokine activity
interleukin-1 receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mmql6:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0