This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03384166_u1
See other GPI GE Assays ›
Gene Symbol:
GPI
Gene Name:
glucose-6-phosphate isomerase
Gene Aliases:
PGI
Chromosome Location:
Chr.6: 44038374 - 44069331 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03384166_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214330.1 NP_999495.1 12 1053 69
GenBank mRNA AK236732.1 - 12 1030 69
AK348401.1 - 12 1092 69
AK389378.1 - 12 1114 69
AK392592.1 - 12 1034 69
AK400763.1 - 2 143 69
X07382.1 - 1053 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
mesoderm formation
glycolytic process
signal transduction
erythrocyte homeostasis
glucose homeostasis
glucose 6-phosphate metabolic process
glucose-6-phosphate isomerase activity
cytokine activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0