This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03384646_u1
See other GPX4 GE Assays ›
Gene Symbol:
GPX4
Gene Name:
glutathione peroxidase 4
Gene Aliases:
PHGPx
Chromosome Location:
Chr.2: 77321568 - 77323931 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03384646_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPX4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214407.1 NP_999572.1 5 - 6 556 75
GenBank mRNA AK232479.1 - 5 - 6 471 75
AK346713.1 - 5 - 6 492 75
AK392406.1 - 5 - 6 530 75
L12743.1 - 5 - 6 556 75
S80257.1 - 4 - 5 381 75
X76009.1 - 467 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
response to oxidative stress
multicellular organism development
spermatogenesis
arachidonic acid metabolic process
lipoxygenase pathway
protein polymerization
oxidation-reduction process
cellular oxidant detoxification
negative regulation of ferroptosis
identical protein binding
phospholipid-hydroperoxide glutathione peroxidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0