This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03385261_u1
See other EEF1A1 GE Assays ›
Gene Symbol:
EEF1A1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
EF1A1, ef1alpha
Chromosome Location:
Chr.1: 92421235 - 92425053 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03385261_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001097418.2 NP_001090887.1 3 345 77
XM_005654402.3 XP_005654459.1 577 77
GenBank mRNA AK236823.1 - 3 346 77
AK236840.1 - 3 345 77
AK239711.1 - 3 345 77
AK347401.1 - 3 345 77
AK391046.1 - 3 345 77
AK391797.1 - 3 346 77
AK393900.1 - 3 345 77
AK393917.1 - 3 376 77
AK394419.1 - 3 345 77
AK394543.1 - 3 433 77
AK396072.1 - 3 345 77
AK396454.1 - 3 345 77
AK397062.1 - 3 345 77
AK397069.1 - 3 345 77
AK397261.1 - 3 344 77
AK397374.1 - 3 510 77
AK397467.1 - 3 350 77
AK397567.1 - 3 345 77
AK397588.1 - 3 346 77
AK397940.1 - 3 347 77
AK399030.1 - 3 347 77
AK399169.1 - 3 348 77
AK399536.1 - 3 345 77
AK399570.1 - 3 348 77
AK399613.1 - 3 348 77
AK399889.1 - 3 346 77
AK400327.1 - 3 345 77
AK400612.1 - 3 346 77
AK400796.1 - 3 345 77
AK400799.1 - 3 345 77
AK400855.1 - 3 345 77
AK400938.1 - 3 345 77
AK400949.1 - 3 349 77
AM040195.1 - 1 110 77
DQ673096.1 - 3 346 77
FM995600.1 - 3 331 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

translation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0