This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03387383_u1
See other AIMP1 GE Assays ›
Gene Symbol:
AIMP1
Gene Name:
aminoacyl tRNA synthetase complex interacting multifunctional protein 1
Gene Aliases:
SCYE1
Chromosome Location:
Chr.8: 115437453 - 115476735 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03387383_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AIMP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114283.1 NP_001107755.1 7 950 106
XM_021100267.1 XP_020955926.1 1088 106
XM_021100268.1 XP_020955927.1 1193 106
XM_021100269.1 XP_020955928.1 1116 106
GenBank mRNA AK392450.1 - 7 995 106
AK400419.1 - 7 106
AY610202.1 - 8 1017 106
EU352799.1 - 7 950 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline