This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03387449_u1
See other LIMA1 GE Assays ›
Gene Symbol:
LIMA1
Gene Name:
LIM domain and actin binding 1
Gene Aliases:
EPLIN
Chromosome Location:
Chr.5: 16074436 - 16177122 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03387449_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIMA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114676.1 NP_001108148.1 11 2250 98
XM_021090869.1 XP_020946528.1 2339 98
XM_021090870.1 XP_020946529.1 2416 98
XM_021090871.1 XP_020946530.1 2508 98
XM_021090873.1 XP_020946532.1 2336 98
XM_021090874.1 XP_020946533.1 2333 98
XM_021090875.1 XP_020946534.1 2390 98
XM_021090876.1 XP_020946535.1 2293 98
XM_021090877.1 XP_020946536.1 2370 98
XM_021090878.1 XP_020946537.1 2456 98
XM_021090879.1 XP_020946538.1 2266 98
GenBank mRNA DQ364062.1 - 11 2250 98
DQ364063.1 - 8 2144 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cholesterol metabolic process
cell migration
intestinal cholesterol absorption
negative regulation of actin filament depolymerization
ruffle organization
cholesterol homeostasis
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0