This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03388941_m1
See other NECTIN2 GE Assays ›
Gene Symbol:
NECTIN2
Gene Name:
nectin cell adhesion molecule 2
Gene Aliases:
PRR2
Chromosome Location:
Chr.6: 51317386 - 51358665 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03388941_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NECTIN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001097494.1 NP_001090963.1 2 - 3 483 149
XM_005653278.2 XP_005653335.1 754 149
XM_013994909.2 XP_013850363.1 754 149
XM_013994914.2 XP_013850368.1 754 149
XM_021093178.1 XP_020948837.1 754 149
GenBank mRNA AK236608.1 - 2 - 3 691 149
AK394295.1 - 2 - 3 680 149
EF195266.1 - 2 - 3 483 149
EU069826.1 - 2 - 3 483 149

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0