This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03390324_m1
See other CREB3L4 GE Assays ›
Gene Symbol:
CREB3L4
Gene Name:
cAMP responsive element binding protein 3 like 4
Gene Aliases:
-
Chromosome Location:
Chr.4: 95716530 - 95723959 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03390324_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CREB3L4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001123102.1 NP_001116574.1 4 - 5 692 107
XM_005663345.3 XP_005663402.1 664 107
XM_005663346.3 XP_005663403.1 1458 107
XM_005663347.3 XP_005663404.1 696 107
GenBank mRNA AK344313.1 - 4 - 5 831 107
EU427542.1 - 4 - 5 692 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline