This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03391409_m1
See other GSTO1 GE Assays ›
Gene Symbol:
GSTO1
Gene Name:
glutathione S-transferase omega 1
Gene Aliases:
GSTO 1-1, SPG-R
Chromosome Location:
Chr.14: 115206434 - 115216624 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03391409_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214050.2 NP_999215.1 4 - 5 537 122
GenBank mRNA AF188838.3 - 4 - 5 540 122
AK235250.1 - 4 - 5 689 122
AK391766.1 - 4 - 5 564 122
AY609655.1 - 4 - 5 686 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

L-ascorbic acid metabolic process
xenobiotic catabolic process
cellular response to arsenic-containing substance
cellular oxidant detoxification
glutathione transferase activity
oxidoreductase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0