This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03392483_u1
See other AKR1A1 GE Assays ›
Gene Symbol:
AKR1A1
Gene Name:
aldo-keto reductase family 1 member A1
Gene Aliases:
ALR1, Akr1a4
Chromosome Location:
Chr.6: 165811245 - 165825168 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03392483_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AKR1A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213890.2 NP_999055.1 7 845 78
XM_021093494.1 XP_020949153.1 1172 78
XM_021093495.1 XP_020949154.1 1361 78
XM_021093497.1 XP_020949156.1 1079 78
XM_021093498.1 XP_020949157.1 3026 78
GenBank mRNA AK348715.1 - 8 923 78
AK389410.1 - 7 919 78
AK390941.1 - 8 923 78
AY609774.1 - 7 880 78
U46064.1 - 7 848 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

L-ascorbic acid biosynthetic process
D-glucuronate catabolic process
oxidation-reduction process
cellular detoxification of aldehyde
retinol dehydrogenase activity
mevaldate reductase (NADPH) activity
allyl-alcohol dehydrogenase activity
L-glucuronate reductase activity
glucuronolactone reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0