This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03394088_g1
See other PPP1R14B GE Assays ›
Gene Symbol:
PPP1R14B
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 14B
Gene Aliases:
PHI-1, pPHI-1
Chromosome Location:
Chr.2: 7865445 - 7867878 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03394088_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R14B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214115.1 NP_999280.1 3 - 4 384 56
GenBank mRNA AK392243.1 - 3 - 4 394 56
AY122324.1 - 3 - 4 384 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s)

negative regulation of phosphoprotein phosphatase activity
regulation of phosphorylation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0