This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03394221_m1
See other PIGR GE Assays ›
Gene Symbol:
PIGR
Gene Name:
polymeric immunoglobulin receptor
Gene Aliases:
-
Chromosome Location:
Chr.9: 67553094 - 67574698 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03394221_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PIGR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214159.1 NP_999324.1 9 - 10 2236 60
XM_021102216.1 XP_020957875.1 2389 60
GenBank mRNA AB032195.2 - 9 - 10 2236 60
AK238185.1 - 7 - 8 1578 60
AK346212.1 - 10 - 11 2333 60
AK392828.1 - 9 - 10 2278 60
AK397936.1 - 10 - 11 2359 60
AK397951.1 - 2 - 3 177 60
AK398076.1 - 6 - 7 1144 60
AK398099.1 - 4 - 5 384 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0