This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03394721_m1
See other IL4R GE Assays ›
Gene Symbol:
IL4R
Gene Name:
interleukin 4 receptor
Gene Aliases:
IL4
Chromosome Location:
Chr.3: 19526343 - 19576792 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03394721_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL4R
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214340.1 NP_999505.1 7 - 8 874 60
XM_021085863.1 XP_020941522.1 1049 60
XM_021085864.1 XP_020941523.1 1052 60
XM_021085865.1 XP_020941524.1 1317 60
XM_021085866.1 XP_020941525.1 1022 60
XM_021085867.1 XP_020941526.1 1057 60
XM_021085868.1 XP_020941527.1 1019 60
XM_021085869.1 XP_020941528.1 1188 60
XM_021085870.1 XP_020941529.1 1006 60
XM_021085871.1 XP_020941530.1 1031 60
XM_021085872.1 XP_020941531.1 1297 60
XM_021085873.1 XP_020941532.1 1001 60
XM_021085874.1 XP_020941533.1 1022 60
XM_021085875.1 XP_020941534.1 1191 60
XM_021085876.1 XP_020941535.1 735 60
GenBank mRNA AK350509.1 - 8 - 9 1002 60
AY266143.1 - 7 - 8 874 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

production of molecular mediator involved in inflammatory response
cytokine-mediated signaling pathway
cytokine receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0