This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03394782_g1
See other PPIA GE Assays ›
Gene Symbol:
PPIA
Gene Name:
peptidylprolyl isomerase A
Gene Aliases:
CYPA
Chromosome Location:
Chr.18: 50563130 - 50566857 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03394782_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPIA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214353.1 NP_999518.1 4 - 5 366 91
XM_013985800.2 XP_013841254.1 470 91
XM_021078519.1 XP_020934178.1 994 91
GenBank mRNA AK231482.1 - 4 - 5 409 91
AY266299.1 - 4 - 5 366 91
AY610237.1 - 4 - 5 384 91
FN401368.1 - 4 - 5 382 91
JX523418.1 - 4 - 5 362 91
JX523419.1 - 4 - 5 362 91
JX523420.1 - 4 - 5 362 91
JX523421.1 - 4 - 5 362 91
JX523422.1 - 4 - 5 362 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Endogenous Controls Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline