This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03394788_m1
See other EPHX1 GE Assays ›
Gene Symbol:
EPHX1
Gene Name:
epoxide hydrolase 1
Gene Aliases:
mEH
Chromosome Location:
Chr.10: 13757489 - 13773071 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03394788_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPHX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214355.1 NP_999520.1 6 - 7 1050 63
GenBank mRNA AB000883.1 - 6 - 7 1050 63
AK232417.1 - 7 - 8 1110 63
AK232523.1 - 7 - 8 1110 63
AK232937.1 - 7 - 8 1141 63
AK233444.1 - 7 - 8 1163 63
AK233660.1 - 7 - 8 1149 63
AK236389.1 - 8 - 9 1304 63
AK238449.1 - 7 - 8 1123 63
AK395509.1 - 7 - 8 1372 63
AK395862.1 - 7 - 8 1256 63
AK396712.1 - 7 - 8 1209 63
AK397552.1 - 7 - 8 1229 63
AK398153.1 - 7 - 8 1169 63
AK398259.1 - 8 - 9 1300 63
AK398297.1 - 7 - 8 1164 63
AK400516.1 - 7 - 8 1154 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

hydrolase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline