This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04246169_m1
See other NQO1 GE Assays ›
Gene Symbol:
NQO1
Gene Name:
NAD(P)H quinone dehydrogenase 1
Gene Aliases:
NADPH-d
Chromosome Location:
Chr.6: 17292962 - 17302593 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04246169_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NQO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159613.1 NP_001153085.1 4 - 5 674 62
GenBank mRNA AK234062.1 - 4 - 5 706 62
AK234977.1 - 3 - 4 540 62
AK351118.1 - 3 - 4 368 62
AK390517.1 - 4 - 5 718 62
AK397043.1 - 4 - 5 558 62
AY610195.1 - 4 - 5 674 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0