This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04246212_g1
See other DGAT2 GE Assays ›
Gene Symbol:
DGAT2
Gene Name:
diacylglycerol O-acyltransferase 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 10032163 - 10066082 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04246212_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DGAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160080.1 NP_001153552.1 5 - 6 721 56
GenBank mRNA AK390770.1 - 5 - 6 552 56
EU684958.1 - 5 - 6 721 56
HQ403606.1 - 5 - 6 558 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0