This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04246348_m1
See other SOCS4 GE Assays ›
Gene Symbol:
SOCS4
Gene Name:
suppressor of cytokine signaling 4
Gene Aliases:
SOCS6
Chromosome Location:
Chr.1: 184379017 - 184398902 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04246348_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SOCS4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162889.1 NP_001156361.1 1 - 2 301 88
GenBank mRNA AK237383.1 - 1 - 2 301 88
FJ971662.1 - 1 - 2 301 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001162889.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

kinase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline