This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04247318_g1
See other PCBD1 GE Assays ›
Gene Symbol:
PCBD1
Gene Name:
pterin-4 alpha-carbinolamine dehydratase 1
Gene Aliases:
-
Chromosome Location:
Chr.14: 73769339 - 73807278 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04247318_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCBD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190219.1 NP_001177148.1 1 - 2 89 80
XM_021071953.1 XP_020927612.1 120 80
XM_021071954.1 XP_020927613.1 120 80
GenBank mRNA AK392722.1 - 1 - 2 94 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001190219.1, XM_021071953.1, XM_021071954.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0