This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04247380_g1
See other HPGD GE Assays ›
Gene Symbol:
HPGD
Gene Name:
15-hydroxyprostaglandin dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.14: 15718261 - 15773534 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04247380_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HPGD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190248.1 NP_001177177.1 1 - 2 96 93
XM_021071974.1 XP_020927633.1 114 93
XM_021071975.1 XP_020927634.1 564 93
XM_021071976.1 XP_020927635.1 564 93
XM_021071977.1 XP_020927636.1 564 93
XM_021071978.1 XP_020927637.1 510 93
GenBank mRNA AB529533.1 - 1 - 2 96 93
AK232290.1 - 1 - 2 147 93
AY609554.1 - 1 - 2 113 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021071974.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f9n9c:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0