This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04247884_m1
See other CPSF6 GE Assays ›
Gene Symbol:
CPSF6
Gene Name:
cleavage and polyadenylation specific factor 6
Gene Aliases:
-
Chromosome Location:
Chr.5: 33525668 - 33559941 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04247884_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CPSF6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195331.1 NP_001182260.1 7 - 8 1332 72
XM_021090899.1 XP_020946558.1 1520 72
XM_021090900.1 XP_020946559.1 1520 72
XM_021090901.1 XP_020946560.1 1520 72
XM_021090902.1 XP_020946561.1 1520 72
XM_021090903.1 XP_020946562.1 1409 72
XM_021090904.1 XP_020946563.1 1409 72
XM_021090905.1 XP_020946564.1 1409 72
GenBank mRNA AK236866.1 - 7 - 8 1358 72
AK391105.1 - 7 - 8 1380 72
GU373705.1 - 7 - 8 1332 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

RNA processing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline