This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04321667_m1
See other PCMT1 GE Assays ›
Gene Symbol:
PCMT1
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase
Gene Aliases:
PCMT, PIMT
Chromosome Location:
Chr.1: 16202859 - 16249879 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04321667_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195780.1 NP_001182709.1 6 - 7 528 61
XM_021067017.1 XP_020922676.1 705 61
XM_021067054.1 XP_020922713.1 696 61
XM_021067107.1 XP_020922766.1 1040 61
GenBank mRNA AF239700.1 - 6 - 7 528 61
AK232514.1 - 6 - 7 696 61
AK392750.1 - 6 - 7 860 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s)

protein methylation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline