This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04321727_m1
See other UBE2I GE Assays ›
Gene Symbol:
UBE2I
Gene Name:
ubiquitin conjugating enzyme E2 I
Gene Aliases:
UBC9
Chromosome Location:
Chr.3: 40556556 - 40586382 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04321727_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2I
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204369.1 NP_001191298.1 6 - 7 541 80
XM_005655131.3 XP_005655188.1 702 80
XM_005655133.3 XP_005655190.1 491 80
XM_021085704.1 XP_020941363.1 735 80
XM_021085705.1 XP_020941364.1 7798 80
XM_021085706.1 XP_020941365.1 550 80
XM_021085707.1 XP_020941366.1 7780 80
GenBank mRNA AK231371.1 - 6 - 7 807 80
AK235702.1 - 7 - 8 629 80
AK239959.1 - 6 - 7 547 80
AK347895.1 - 6 - 7 541 80
AK396107.1 - 6 - 7 539 80
AY610452.1 - 6 - 7 493 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0