This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04321787_g1
See other POLR2I GE Assays ›
Gene Symbol:
POLR2I
Gene Name:
RNA polymerase II subunit I
Gene Aliases:
RPB14.5
Chromosome Location:
Chr.6: 45480785 - 45482318 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04321787_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR2I
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205404.1 NP_001192333.1 2 - 3 480 62
NM_001205406.1 NP_001192335.1 1 - 2 201 62
GenBank mRNA AK234376.1 - 2 - 3 142 62
AK400001.1 - 1 - 2 201 62
EW379062.2 - 1 - 2 220 62
FD595099.1 - 2 - 3 480 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
mRNA cleavage
nucleic acid binding
DNA-directed RNA polymerase activity
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0