This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04322612_g1
See other CIRBP GE Assays ›
Gene Symbol:
CIRBP
Gene Name:
cold inducible RNA binding protein
Gene Aliases:
CIRP
Chromosome Location:
Chr.2: 77185295 - 77194373 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04322612_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CIRBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001246197.1 NP_001233126.1 2 - 3 195 66
XM_005661375.3 XP_005661432.1 238 66
XM_005661377.3 XP_005661434.1 238 66
XM_005661378.3 XP_005661435.1 266 66
XM_005661379.3 XP_005661436.1 238 66
XM_005661381.3 XP_005661438.1 238 66
XM_005661382.3 XP_005661439.1 238 66
XM_013994815.2 XP_013850269.1 238 66
XM_021079351.1 XP_020935010.1 238 66
XM_021079362.1 XP_020935021.1 266 66
XM_021079369.1 XP_020935028.1 266 66
XM_021079372.1 XP_020935031.1 238 66
XM_021079384.1 XP_020935043.1 238 66
XM_021079389.1 XP_020935048.1 238 66
XM_021079392.1 XP_020935051.1 266 66
XM_021079398.1 XP_020935057.1 277 66
XM_021079405.1 XP_020935064.1 266 66
XM_021079410.1 XP_020935069.1 238 66
XM_021079413.1 XP_020935072.1 238 66
GenBank mRNA AK231744.1 - 2 - 3 183 66
AK234205.1 - 2 - 3 189 66
AK235139.1 - 2 - 3 216 66
AK394938.1 - 2 - 3 219 66
AK400883.1 - 2 - 3 185 66
HQ908794.1 - 2 - 3 195 66
HQ908795.1 - 2 - 3 195 66
HQ908796.1 - 2 - 3 195 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0