This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04322971_m1
See other EWSR1 GE Assays ›
Gene Symbol:
EWSR1
Gene Name:
EWS RNA binding protein 1
Gene Aliases:
EWS
Chromosome Location:
Chr.14: 46408636 - 46439357 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04322971_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EWSR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252048.1 NP_001238977.1 5 - 6 579 68
XM_005657339.3 XP_005657396.1 684 68
XM_005657340.3 XP_005657397.1 684 68
XM_005657341.3 XP_005657398.1 666 68
XM_005657342.3 XP_005657399.1 666 68
XM_013990377.2 XP_013845831.1 684 68
XM_013990378.2 XP_013845832.1 684 68
XM_021072000.1 XP_020927659.1 684 68
XM_021072001.1 XP_020927660.1 684 68
XM_021072003.1 XP_020927662.1 666 68
XM_021072004.1 XP_020927663.1 666 68
XM_021072005.1 XP_020927664.1 666 68
GenBank mRNA AK232912.1 - 6 - 7 643 68
JF940527.1 - 5 - 6 579 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0