This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04323377_m1
See other ADGRG1 GE Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
GPR56
Chromosome Location:
Chr.6: 19523618 - 19570891 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04323377_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243056.1 NP_001229985.1 5 - 6 919 65
XM_021093328.1 XP_020948987.1 1026 65
XM_021093330.1 XP_020948989.1 1125 65
XM_021093331.1 XP_020948990.1 871 65
XM_021093332.1 XP_020948991.1 847 65
XM_021093333.1 XP_020948992.1 1321 65
XM_021093334.1 XP_020948993.1 1043 65
XM_021093335.1 XP_020948994.1 901 65
XM_021093336.1 XP_020948995.1 2457 65
XM_021093337.1 XP_020948996.1 1080 65
XM_021093338.1 XP_020948997.1 995 65
GenBank mRNA AK348726.1 - 5 - 6 919 65
AK349595.1 - 5 - 6 1138 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0