Assay Details
| Gene Symbol: MMGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243208.1 | NP_001230137.1 | 1 - 2 | 147 | 72 | |
| GenBank mRNA | AK344491.1 | - | 1 - 2 | 116 | 72 | |
| FS668658.1 | - | 1 - 2 | 148 | 72 | ||
More Information
Set Membership: |
|

MMGT1
Genomic Map