This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04323895_m1
See other PLA2G12B GE Assays ›
Gene Symbol:
PLA2G12B
Gene Name:
phospholipase A2 group XIIB
Gene Aliases:
-
Chromosome Location:
Chr.14: 75754525 - 75778933 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04323895_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLA2G12B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243338.1 NP_001230267.1 1 - 2 303 94
XM_005671053.3 XP_005671110.1 371 94
XM_005671054.3 XP_005671111.1 374 94
XM_005671055.3 XP_005671112.1 374 94
GenBank mRNA AK345294.1 - 1 - 2 364 94
AK346162.1 - 1 - 2 364 94
EW113750.2 - 1 - 2 328 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0