This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04324354_m1
See other EIF4H GE Assays ›
Gene Symbol:
EIF4H
Gene Name:
eukaryotic translation initiation factor 4H
Gene Aliases:
-
Chromosome Location:
Chr.3: 11321486 - 11348473 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04324354_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF4H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243447.1 NP_001230376.1 1 - 2 118 75
NM_001243448.1 NP_001230377.1 1 - 2 118 75
GenBank mRNA AK348471.1 - 1 - 2 118 75
AK390998.1 - 1 - 2 61 75
AK396280.1 - 1 - 2 200 75
AK399888.1 - 1 - 2 88 75
BP150090.1 - 1 - 2 87 75
BP462018.1 - 1 - 2 119 75
CV872394.1 - 1 - 2 237 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0