This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04324461_m1
See other DUSP13 GE Assays ›
Gene Symbol:
DUSP13
Gene Name:
dual specificity phosphatase 13
Gene Aliases:
-
Chromosome Location:
Chr.14: 77660124 - 77677713 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04324461_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUSP13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243472.1 NP_001230401.1 3 - 4 663 62
XM_005671056.3 XP_005671113.2 896 62
XM_013983312.2 XP_013838766.1 826 62
XM_013983314.2 XP_013838768.1 564 62
XM_021071924.1 XP_020927583.1 2633 62
XM_021071925.1 XP_020927584.1 3116 62
XM_021071926.1 XP_020927585.1 789 62
XM_021071927.1 XP_020927586.1 577 62
GenBank mRNA AK346500.1 - 3 - 4 663 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0