This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04325019_m1
See other GSTZ1 GE Assays ›
Gene Symbol:
GSTZ1
Gene Name:
glutathione S-transferase zeta 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 100392217 - 100404025 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04325019_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTZ1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243638.1 NP_001230567.1 7 - 8 599 99
XM_005656430.3 XP_005656487.1 564 99
XM_013989192.2 XP_013844646.1 593 99
XM_013989193.2 XP_013844647.1 519 99
XM_013989194.2 XP_013844648.1 545 99
XM_021098266.1 XP_020953925.1 569 99
XM_021098267.1 XP_020953926.1 2297 99
GenBank mRNA FD609421.1 - 7 - 8 601 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0