This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04325125_g1
See other PAQR6 GE Assays ›
Gene Symbol:
PAQR6
Gene Name:
progestin and adipoQ receptor family member 6
Gene Aliases:
-
Chromosome Location:
Chr.4: 93802721 - 93807624 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04325125_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PAQR6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243668.1 NP_001230597.1 3 - 4 237 93
XM_005663288.3 XP_005663345.1 807 93
XM_005663289.3 XP_005663346.1 377 93
XM_005663290.3 XP_005663347.1 257 93
XM_013996967.2 XP_013852421.1 859 93
XM_013996968.2 XP_013852422.1 813 93
XM_013996969.2 XP_013852423.1 866 93
XM_013996970.2 XP_013852424.1 866 93
XM_013996971.2 XP_013852425.1 807 93
XM_013996974.2 XP_013852428.1 290 93
XM_013996975.2 XP_013852429.1 863 93
XM_021088598.1 XP_020944257.1 862 93
XM_021088599.1 XP_020944258.1 501 93
XM_021088600.1 XP_020944259.1 810 93
XM_021088601.1 XP_020944260.1 380 93
XM_021088602.1 XP_020944261.1 868 93
GenBank mRNA AK389884.1 - 3 - 4 295 93
EU649704.1 - 2 - 3 186 93
FS665696.1 - 3 - 4 238 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0