Assay Details
| Gene Symbol: KRT74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243814.1 | NP_001230743.1 | 3 - 4 | 815 | 67 | |
| GenBank mRNA | AK349684.1 | - | 3 - 4 | 815 | 67 | |
More Information
Set Membership: |
|

KRT74
Genomic Map