This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04325909_m1
See other TMEM183A GE Assays ›
Gene Symbol:
TMEM183A
Gene Name:
transmembrane protein 183A
Gene Aliases:
-
Chromosome Location:
Chr.9: 113713046 - 113726645 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04325909_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM183A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243927.1 NP_001230856.1 6 - 7 866 77
XM_021102013.1 XP_020957672.1 898 77
GenBank mRNA AK234299.1 - 6 - 7 866 77
AK235615.1 - 6 - 7 866 77
AK396401.1 - 6 - 7 892 77
AK399027.1 - 6 - 7 909 77
AY609482.1 - 6 - 7 863 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0