This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04327630_m1
See other GLUD1 GE Assays ›
Gene Symbol:
GLUD1
Gene Name:
glutamate dehydrogenase 1
Gene Aliases:
GLUD, MP50
Chromosome Location:
Chr.14: 87968850 - 88005375 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04327630_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLUD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244501.2 NP_001231430.1 4 - 5 775 55
GenBank mRNA AK234901.1 - 4 - 5 842 55
AK235921.1 - 4 - 5 450 55
AK392294.1 - 4 - 5 775 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

glutamine metabolic process
oxidation-reduction process
tricarboxylic acid metabolic process
glutamate dehydrogenase [NAD(P)+] activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0