This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04327880_m1
See other H2AFY GE Assays ›
Gene Symbol:
H2AFY
Gene Name:
H2A histone family, member Y
Gene Aliases:
-
Chromosome Location:
Chr.2: 137535128 - 137623886 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04327880_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H2AFY
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244562.1 NP_001231491.1 8 - 9 1099 57
NM_001244563.1 NP_001231492.1 8 - 9 1149 57
XM_021078328.1 XP_020933987.1 1196 57
XM_021078334.1 XP_020933993.1 1004 57
XM_021078345.1 XP_020934004.1 1098 57
XM_021078354.1 XP_020934013.1 1193 57
XM_021078358.1 XP_020934017.1 6982 57
XM_021078365.1 XP_020934024.1 1187 57
XM_021078375.1 XP_020934034.1 1086 57
XM_021078384.1 XP_020934043.1 6973 57
GenBank mRNA AK235350.1 - 8 - 9 1184 57
AK350276.1 - 8 - 9 1187 57
AK351941.1 - 8 - 9 1099 57
AK389385.1 - 8 - 9 1187 57
AK394143.1 - 8 - 9 1187 57
AK395913.1 - 8 - 9 1158 57
AK396498.1 - 8 - 9 1193 57
AK398380.1 - 8 - 9 1099 57
AK398405.1 - 8 - 9 1158 57
AK398693.1 - 8 - 9 1201 57
BW973982.1 - 4 - 5 436 57
EW242020.2 - 2 - 3 238 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

histone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5vr65:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0