This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04328362_m1
See other VAMP4 GE Assays ›
Gene Symbol:
VAMP4
Gene Name:
vesicle associated membrane protein 4
Gene Aliases:
-
Chromosome Location:
Chr.9: 114138350 - 114174996 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04328362_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VAMP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244707.1 NP_001231636.1 4 - 5 370 79
XM_005653719.3 XP_005653776.1 398 79
XM_013977625.2 XP_013833079.1 443 79
XM_021102024.1 XP_020957683.1 303 79
GenBank mRNA AK237186.1 - 4 - 5 370 79
AK395657.1 - 4 - 5 379 79
AY609450.1 - 3 - 4 211 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0