This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04345017_m1
See other RBBP7 GE Assays ›
Gene Symbol:
RBBP7
Gene Name:
RB binding protein 7, chromatin remodeling factor
Gene Aliases:
-
Chromosome Location:
Chr.X: 13293278 - 13317074 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04345017_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBBP7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003134956.6 XP_003135004.1 1112 77
XM_003360236.5 XP_003360284.1 1099 77
XM_005673458.3 XP_005673515.1 883 77
XM_005673459.2 XP_005673516.1 1042 77
GenBank mRNA AK233215.1 - 6 - 7 1038 77
AK234790.1 - 6 - 7 1041 77
AK348165.1 - 6 - 7 951 77
AK394905.1 - 6 - 7 1044 77
AK396052.1 - 6 - 7 1068 77
AY609492.1 - 6 - 7 1018 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-x666z:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline