This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04652439_sH
See other MIR107 GE Assays ›
Gene Symbol:
MIR107
Gene Name:
microRNA mir-107
Gene Aliases:
MIRN107, ssc-mir-107
Chromosome Location:
Chr.14: 101420743 - 101420829 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04652439_sH
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR107
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031539.1 1 46 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream pantothenate kinase 1 PANK1 Sense Chr. 14 - 101410139 - 101512498 on NCBI Build Sscrofa11.1
Downstream kinesin family member 20B KIF20B 91.9kb Anti-sense Chr. 14 - 101512685 - 101594703 on NCBI Build Sscrofa11.1

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-695c48667f-vfqgd:80/100.66.73.211:80.
git-commit: c086736f0d622c41bbf3ea4d71b1c2b8619a5b9d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.35.0-Offline