This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04953112_m1
See other MLLT11 GE Assays ›
Gene Symbol:
MLLT11
Gene Name:
MLLT11 transcription factor 7 cofactor
Gene Aliases:
-
Chromosome Location:
Chr.4: 98165990 - 98171674 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04953112_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MLLT11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244607.1 NP_001231536.1 1 - 2 154 76
GenBank mRNA AK392540.1 - 1 - 2 155 76
FS666254.1 - 1 - 2 154 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001244607.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s2cfr:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0