This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04954766_g1
See other GSTK1 GE Assays ›
Gene Symbol:
GSTK1
Gene Name:
glutathione S-transferase kappa 1
Gene Aliases:
-
Chromosome Location:
Chr.18: 6992623 - 6999068 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04954766_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001315574.1 NP_001302503.1 7 - 8 772 61
GenBank mRNA AK232526.1 - 7 - 8 731 61
AK234199.1 - 7 - 8 731 61
AK346267.1 - 7 - 8 731 61
AK348138.1 - 7 - 8 731 61
AK391176.1 - 7 - 8 782 61
AK399796.1 - 7 - 8 733 61
AY609983.1 - 7 - 8 714 61
KP735779.1 - 7 - 8 772 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

transferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0