This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04954785_m1
See other ALDH3B1 GE Assays ›
Gene Symbol:
ALDH3B1
Gene Name:
aldehyde dehydrogenase 3 family member B1
Gene Aliases:
-
Chromosome Location:
Chr.2: 4868561 - 4889179 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04954785_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH3B1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001315593.1 NP_001302522.1 3 - 4 332 72
XM_021080123.1 XP_020935782.1 390 72
XM_021080124.1 XP_020935783.1 537 72
XM_021080126.1 XP_020935785.1 409 72
XM_021080130.1 XP_020935789.1 311 72
GenBank mRNA AK240330.1 - 3 - 4 341 72
KP735777.1 - 3 - 4 332 72
KP735784.1 - 3 - 4 332 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021080130.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0