This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04954832_m1
See other SLCO1B3 GE Assays ›
Gene Symbol:
SLCO1B3
Gene Name:
solute carrier organic anion transporter family member 1B3
Gene Aliases:
Oatp1b4
Chromosome Location:
Chr.5: 52137454 - 52206236 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04954832_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO1B3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001315607.1 NP_001302536.1 7 - 8 966 151
XM_005655659.3 XP_005655716.1 1094 151
XM_021091087.1 XP_020946746.1 1128 151
XM_021091089.1 XP_020946748.1 1260 151
XM_021091090.1 XP_020946749.1 1265 151
XM_021091091.1 XP_020946750.1 950 151
GenBank mRNA AK394342.1 - 5 - 6 735 151
LN833582.1 - 7 - 8 966 151

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0