This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04954918_m1
See other UGT8 GE Assays ›
Gene Symbol:
UGT8
Gene Name:
UDP glycosyltransferase 8
Gene Aliases:
-
Chromosome Location:
Chr.8: 108233134 - 108315284 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04954918_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UGT8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001315662.1 NP_001302591.1 4 - 5 1263 87
XM_021100329.1 XP_020955988.1 1490 87
XM_021100330.1 XP_020955989.1 1853 87
XM_021100331.1 XP_020955990.1 1894 87
XM_021100332.1 XP_020955991.1 1285 87
GenBank mRNA JQ650526.1 - 4 - 5 1263 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0