This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04955200_s1
See other GPR6 GE Assays ›
Gene Symbol:
GPR6 LOC110255904
Gene Name:
G protein-coupled receptor 6
G-protein coupled receptor 6-like
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 75979360 - 75982036 on Build Sscrofa11.1
Chromosome Location:
Chr.12: 41202560 - 41204697 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
-
Assay ID Ss04955200_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001315817.1 NP_001302746.1 1 723 98
XM_003121354.4 XP_003121402.2 873 98
GenBank mRNA HM777010.1 - 1 723 98
Gene Symbol: LOC110255904
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021066310.1 XP_020921969.1 775 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0