This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04955671_mH
See other LIPE GE Assays ›
Gene Symbol:
LIPE
Gene Name:
lipase E, hormone sensitive type
Gene Aliases:
HSL, REH
Chromosome Location:
Chr.6: 49543671 - 49560126 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04955671_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214315.3 NP_999480.2 8 - 9 1710 65
XM_013988600.2 XP_013844054.2 1845 65
XM_013988601.2 XP_013844055.1 2237 65
XM_013988602.2 XP_013844056.1 1776 65
XM_021093574.1 XP_020949233.1 1849 65
GenBank mRNA AJ000482.1 - 7 - 8 2074 65
AY556478.1 - 7 - 8 1685 65
AY556479.1 - 7 - 8 1701 65
AY559451.1 - 7 - 8 1684 65
AY686758.1 - 7 - 8 1695 65
AY686759.1 - 7 - 8 1695 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

transcription initiation from RNA polymerase I promoter
termination of RNA polymerase I transcription
cholesterol metabolic process
lipid catabolic process
triglyceride catabolic process
retinol metabolic process
diacylglycerol catabolic process
ether lipid metabolic process
sterol esterase activity
triglyceride lipase activity
hormone-sensitive lipase activity
rRNA primary transcript binding
acylglycerol lipase activity
retinyl-palmitate esterase activity
1,3-diacylglycerol acylhydrolase activity
1,2-diacylglycerol acylhydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0