This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04956045_s1
See other IFIT2 GE Assays ›
Gene Symbol:
IFIT2
Gene Name:
interferon induced protein with tetratricopeptide repeats 2
Gene Aliases:
G10P2, GARG-39, IFI-54, IFI-54K, IFI54, IFIT-2, ISG-54K, ISG54, cig42
Chromosome Location:
Chr.14: 101193421 - 101204292 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04956045_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IFIT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001315658.1 NP_001302587.1 2 1326 104
XM_005671264.3 XP_005671321.1 1587 104
GenBank mRNA AK230663.1 - 2 1388 104
JN621782.1 - 2 1326 104
JX070559.1 - 2 1382 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline